Learn More
Cayman Chemical Rivaroxaban-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1132681-38-9, 1 mg
SDP
Catalog No. 502964135
Encompass_Preferred
An internal standard for the quantification of rivaroxaban by GC- or LC-MS
Catalog No. 50-296-4135 Supplier Cayman Chemical Supplier No. 225631 MG
An internal standard for the quantification of rivaroxaban by GC- or LC-MS