Learn More
Cayman Chemical RIvaroxabn-d4 500ug
SDP
Catalog No. 502986272
Encompass_Preferred
An internal standard for the quantification of rivaroxaban by GC- or LC-MS
Catalog No. 50-298-6272 Supplier Cayman Chemical Supplier No. 22563500 UG
An internal standard for the quantification of rivaroxaban by GC- or LC-MS