Learn More
Selleck Chemical LLC Rivastigmine S4687-20mg
SDP
Catalog No. 503108180
Encompass_Preferred
Rivastigmine (SDZ-ENA 713 Exelon) is a cholinesterase inhibitor with IC50 of 5 5 M It inhibits acetylcholinesterase (IC50 4 15 M) and butyrylcholinesterase (IC50 37 nM)
Catalog No. 50-310-8180 Supplier Selleck Chemical LLC Supplier No. S468720MG
Rivastigmine (SDZ-ENA 713 Exelon) is a cholinesterase inhibitor with IC50 of 5 5 M It inhibits acetylcholinesterase (IC50 4 15 M) and butyrylcholinesterase (IC50 37 nM)