Learn More
Medchemexpress LLC Rivastigmine-d4 tartrate | 129101-54-8 | 404.45 | C18H24D4N2O8 | 1 MG
SDP
Catalog No. 5000284312
Encompass_Preferred
Rivastigmine-d4 tartrate is a deuterium-labeled analog of rivastigmine tartrate supplied for research use. It is a cholinesterase inhibitor used in studies of neurodegenerative diseases and is suitable as an isotopic internal standard or tracer in analytical and pharmacokinetic workflows.
  • Deuterium-labeled for use as an internal standard and tracer in analytical studies.
  • Used to study cholinesterase inhibition relevant to Alzheimer's and Parkinson's research.
  • Blood-brain barrier permeable analog appropriate for in vitro and in vivo models.
  • High solubility in water (approx. 50 mg/mL) facilitates stock solution preparation.
  • Provided in a small pack size suitable for discovery and analytical workflows.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-84312 Supplier Medchemexpress LLC Supplier No. HYW6543351MG
Rivastigmine-d4 tartrate is a deuterium-labeled analog of rivastigmine tartrate supplied for research use. It is a cholinesterase inhibitor used in studies of neurodegenerative diseases and is suitable as an isotopic internal standard or tracer in analytical and pharmacokinetic workflows.
  • Deuterium-labeled for use as an internal standard and tracer in analytical studies.
  • Used to study cholinesterase inhibition relevant to Alzheimer's and Parkinson's research.
  • Blood-brain barrier permeable analog appropriate for in vitro and in vivo models.
  • High solubility in water (approx. 50 mg/mL) facilitates stock solution preparation.
  • Provided in a small pack size suitable for discovery and analytical workflows.
  • For research use only; not for human or clinical use.