Learn More
Rivastigmine Related Compound F Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 1346602-84-3 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | C12H15 NO2 |
| Synonym | 3-Vinylphenyl ethyl(methyl)carbamate; 3-Vinylphenyl ethyl(methyl)carbamate |
| Type | Certified Reference Material |
| Quantity | 50 mg |
| Formula Weight | 205.25 |
| Grade | Certified Reference Material |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.