Learn More
Apexbio Technology LLC Rivastigmine Tartrate 129101-54-8 50mg
SDP
Catalog No. 5000095182
Encompass_Preferred
Rivastigmine tartrate (CAS 129101-54-8) is a reversible inhibitor of both acetylcholinesterase and butyrylcholinesterase enzymes responsible for the hydrolysis of acetylcholine in the central nervous system By inhibiting these enzymes rivastigmine enhances cholinergic neurotransmission and increases synaptic acetylcholine availability This compound is widely utilized in studies related to cholinergic dysfunction including models of mild to moderate Alzheimer s disease and Parkinson s disease-associated dementia providing a tool for investigating cholinergic signaling pathways and potential therapeutic interventions in neurodegenerative disorders
Catalog No. 50-000-95182 Supplier Apexbio Technology LLC Supplier No. A851550
Rivastigmine tartrate (CAS 129101-54-8) is a reversible inhibitor of both acetylcholinesterase and butyrylcholinesterase enzymes responsible for the hydrolysis of acetylcholine in the central nervous system By inhibiting these enzymes rivastigmine enhances cholinergic neurotransmission and increases synaptic acetylcholine availability This compound is widely utilized in studies related to cholinergic dysfunction including models of mild to moderate Alzheimer s disease and Parkinson s disease-associated dementia providing a tool for investigating cholinergic signaling pathways and potential therapeutic interventions in neurodegenerative disorders