Learn More
Medchemexpress LLC Rivastigmine (tartrate) | 129101-54-8 | MFCD03700731 | 99.03% | 400.42 | 50 MG
SDP
Catalog No. 5000343479
Encompass_Preferred
Rivastigmine tartrate is an orally active and potent cholinesterase (ChE) inhibitor. It inhibits butyrylcholinesterase (BChE) and acetylcholinesterase (AChE). This compound can pass the blood-brain barrier (BBB) and is used for the research of mild to moderate dementia of the Alzheimer's type and dementia due to Parkinson's disease.
  • Orally active and potent cholinesterase (ChE) inhibitor
  • Inhibits butyrylcholinesterase (BChE) and acetylcholinesterase (AChE)
  • Can pass the blood-brain barrier (BBB)
  • Used for research of mild to moderate dementia of the Alzheimer's type and dementia due to Parkinson's disease
  • Reduces LPS-induced TNF-α and IL-6 in vitro
  • Improved behavioral impairments caused by Aluminum in male Wistar albino rats
  • Reduced IL-6 concentration in BALB/c OlaHsd male mice with acute colitis
Catalog No. 50-003-43479 Supplier Medchemexpress LLC Supplier No. HY11017200MG
Rivastigmine tartrate is an orally active and potent cholinesterase (ChE) inhibitor. It inhibits butyrylcholinesterase (BChE) and acetylcholinesterase (AChE). This compound can pass the blood-brain barrier (BBB) and is used for the research of mild to moderate dementia of the Alzheimer's type and dementia due to Parkinson's disease.
  • Orally active and potent cholinesterase (ChE) inhibitor
  • Inhibits butyrylcholinesterase (BChE) and acetylcholinesterase (AChE)
  • Can pass the blood-brain barrier (BBB)
  • Used for research of mild to moderate dementia of the Alzheimer's type and dementia due to Parkinson's disease
  • Reduces LPS-induced TNF-α and IL-6 in vitro
  • Improved behavioral impairments caused by Aluminum in male Wistar albino rats
  • Reduced IL-6 concentration in BALB/c OlaHsd male mice with acute colitis