Learn More
Selleck Chemical LLC Rivastigmine Tartrate S2087-50mg
SDP
Catalog No. 503110016
Encompass_Preferred
Rivastigmine Tartrate (ENA 713) is a cholinesterase inhibitor with IC50 of 5 5 M and used as a parasympathomimetic or cholinergic agent for the treatment of mild to moderate Alzheimer disease
Catalog No. 50-311-0016 Supplier Selleck Chemical LLC Supplier No. S208750MG
Rivastigmine Tartrate (ENA 713) is a cholinesterase inhibitor with IC50 of 5 5 M and used as a parasympathomimetic or cholinergic agent for the treatment of mild to moderate Alzheimer disease