Learn More
Grainger Milwaukee Tool M12 Rivet Tool Kit, 12 V DC, 2,000 lbf, 2-Battery Kit
Catalog No. 502759158
Encompass_Preferred
This cordless rivet tool provides a fast and efficient solution for professional fastening applications. Designed for versatility, the tool holds rivets in any orientation and significantly reduces muscle effort compared to manual alternatives. It is powered by the M12 battery system and is compatible with various material types and rivet diameters.
  • Delivers 2,000 lbf of pulling force
  • Compatible with 3/32 in., 1/8 in., 5/32 in., and 3/16 in. diameter rivets
  • Features a 0.8 in. stroke length for consistent performance
  • Includes two 1.5 Ah batteries and a battery charger
  • Maintains high productivity, completing up to 450 1/8 in. steel rivets per charge
  • Ergonomic design for improved force reduction and ease of use
  • Equipped with a mandrel collector for cleaner operation
  • Includes a carrying case for storage and transport
Catalog No. 50-275-9158 Supplier Grainger Supplier No. 165FV5
This cordless rivet tool provides a fast and efficient solution for professional fastening applications. Designed for versatility, the tool holds rivets in any orientation and significantly reduces muscle effort compared to manual alternatives. It is powered by the M12 battery system and is compatible with various material types and rivet diameters.
  • Delivers 2,000 lbf of pulling force
  • Compatible with 3/32 in., 1/8 in., 5/32 in., and 3/16 in. diameter rivets
  • Features a 0.8 in. stroke length for consistent performance
  • Includes two 1.5 Ah batteries and a battery charger
  • Maintains high productivity, completing up to 450 1/8 in. steel rivets per charge
  • Ergonomic design for improved force reduction and ease of use
  • Equipped with a mandrel collector for cleaner operation
  • Includes a carrying case for storage and transport