Learn More
Medchemexpress LLC HY-16559 5mg Medchemexpress, Riviciclib hydrochloride CAS:920113-03-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY165595MG
Encompass_Preferred
Medchemexpress, HY-16559 5mg Riviciclib HCl CAS:920113-03-7 Riviciclib hydrochloride (P276-00) is a potent cyclin-dependent kinase (CDK) inhibitor, which inhibits CDK9-cyclinT1, CDK4-cyclin D1, and CDK1-cyclinB with IC50s of 20 nM, 63 nM, and 79 nM, respectively.Riviciclib hydrochloride (P276-00) shows antitumor activity on cisplatin-resistant cells[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9974