Learn More
Medchemexpress LLC Riviciclib hydrochloride | 920113-03-7 | 99.08% | 438.30 | 25 MG
SDP
Catalog No. 5000393013
Encompass_Preferred
Riviciclib hydrochloride is a potent cyclin-dependent kinase (CDK) inhibitor. It inhibits CDK9-cyclinT1, CDK4-cyclin D1, and CDK1-cyclinB with IC50s of 20 nM, 63 nM, and 79 nM, respectively. This compound shows antitumor activity on cisplatin-resistant cells.
  • IC50 & Target information available
  • In vitro studies show activity in various cancer cell lines
  • In vivo studies demonstrate significant inhibition of tumor growth
Catalog No. 50-003-93013 Supplier Medchemexpress LLC Supplier No. 25MGHY16559
Riviciclib hydrochloride is a potent cyclin-dependent kinase (CDK) inhibitor. It inhibits CDK9-cyclinT1, CDK4-cyclin D1, and CDK1-cyclinB with IC50s of 20 nM, 63 nM, and 79 nM, respectively. This compound shows antitumor activity on cisplatin-resistant cells.
  • IC50 & Target information available
  • In vitro studies show activity in various cancer cell lines
  • In vivo studies demonstrate significant inhibition of tumor growth