Learn More
Medchemexpress LLC Riviciclib hydrochloride | 920113-03-7 | 99.1% | 438.30 | 100 MG
SDP
Catalog No. 5000391124
Encompass_Preferred
Riviciclib hydrochloride is a potent cyclin-dependent kinase (CDK) inhibitor. It inhibits CDK9-cyclinT1, CDK4-cyclin D1, and CDK1-cyclinB with IC50s of 20 nM, 63 nM, and 79 nM, respectively, and shows antitumor activity on cisplatin-resistant cells.
  • Potent cyclin-dependent kinase (CDK) inhibitor.
  • Inhibits CDK9-cyclinT1 (IC50: 20 nM), CDK4-cyclin D1 (IC50: 63 nM), and CDK1-cyclinB (IC50: 79 nM).
  • Shows antitumor activity on cisplatin-resistant cells.
  • Induces G1-G2 arrest and apoptosis in promyelocytic leukemia cells.
  • Reduces cyclin D1, Cdk4, and Rb levels in H-460 cells.
  • Active in various human cancer cell lines including colon carcinoma, osteosarcoma, cervical carcinoma, and bladder carcinoma cells.
  • Significant in vivo efficacy in tumor models, inhibiting growth in human colon carcinoma HCT-116 xenograft and H-460 tumor xenograft in mice.
Catalog No. 50-003-91124 Supplier Medchemexpress LLC Supplier No. HY16559100MG
Riviciclib hydrochloride is a potent cyclin-dependent kinase (CDK) inhibitor. It inhibits CDK9-cyclinT1, CDK4-cyclin D1, and CDK1-cyclinB with IC50s of 20 nM, 63 nM, and 79 nM, respectively, and shows antitumor activity on cisplatin-resistant cells.
  • Potent cyclin-dependent kinase (CDK) inhibitor.
  • Inhibits CDK9-cyclinT1 (IC50: 20 nM), CDK4-cyclin D1 (IC50: 63 nM), and CDK1-cyclinB (IC50: 79 nM).
  • Shows antitumor activity on cisplatin-resistant cells.
  • Induces G1-G2 arrest and apoptosis in promyelocytic leukemia cells.
  • Reduces cyclin D1, Cdk4, and Rb levels in H-460 cells.
  • Active in various human cancer cell lines including colon carcinoma, osteosarcoma, cervical carcinoma, and bladder carcinoma cells.
  • Significant in vivo efficacy in tumor models, inhibiting growth in human colon carcinoma HCT-116 xenograft and H-460 tumor xenograft in mice.