Learn More
Medchemexpress LLC Riviciclib hydrochloride | 920113-03-7 | 99.1% | 438.30 | C21H21Cl2NO5 | 10 MG
SDP
Catalog No. 5000288921
Encompass_Preferred
Riviciclib hydrochloride is a potent cyclin-dependent kinase inhibitor provided for preclinical research. It inhibits multiple CDK complexes, induces apoptosis and cell cycle arrest in cancer cell lines, and has shown antitumor activity in xenograft models. Supplied as an off-white to light yellow solid with high reported purity, it is intended for laboratory studies of cell cycle regulation and oncology-related mechanisms.
  • Potent inhibition of CDK9, CDK4, and CDK1 with low-nanomolar to sub-micromolar IC50s.
  • Induces apoptosis and G1 cell cycle arrest in susceptible cell lines.
  • Demonstrated tumor growth inhibition in preclinical xenograft studies.
  • High purity suitable for research use applications.
  • Packaged in a small 10 mg quantity for bench-scale experiments.
Catalog No. 50-002-88921 Supplier Medchemexpress LLC Supplier No. HY1655910MG
Riviciclib hydrochloride is a potent cyclin-dependent kinase inhibitor provided for preclinical research. It inhibits multiple CDK complexes, induces apoptosis and cell cycle arrest in cancer cell lines, and has shown antitumor activity in xenograft models. Supplied as an off-white to light yellow solid with high reported purity, it is intended for laboratory studies of cell cycle regulation and oncology-related mechanisms.
  • Potent inhibition of CDK9, CDK4, and CDK1 with low-nanomolar to sub-micromolar IC50s.
  • Induces apoptosis and G1 cell cycle arrest in susceptible cell lines.
  • Demonstrated tumor growth inhibition in preclinical xenograft studies.
  • High purity suitable for research use applications.
  • Packaged in a small 10 mg quantity for bench-scale experiments.