Learn More
Apexbio Technology LLC Rizatriptan Benzoate 145202-66-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000095322
Encompass_Preferred
Rizatriptan Benzoate (CAS 145202-66-0) is a synthetic small molecule structurally related to tryptamine that acts as a selective agonist at serotonin 5-HT1D receptors By targeting these G protein-coupled receptors Rizatriptan Benzoate modulates the release of vasoactive neuropeptides and inhibits neurogenic inflammation and vasodilation associated with cranial blood vessels This pharmacological profile has made it a valuable tool for investigating serotonergic signaling pathways receptor pharmacodynamics and migraine pathophysiology in preclinical research settings
Catalog No. 50-000-95322 Supplier Apexbio Technology LLC Supplier No. A85165.1
Rizatriptan Benzoate (CAS 145202-66-0) is a synthetic small molecule structurally related to tryptamine that acts as a selective agonist at serotonin 5-HT1D receptors By targeting these G protein-coupled receptors Rizatriptan Benzoate modulates the release of vasoactive neuropeptides and inhibits neurogenic inflammation and vasodilation associated with cranial blood vessels This pharmacological profile has made it a valuable tool for investigating serotonergic signaling pathways receptor pharmacodynamics and migraine pathophysiology in preclinical research settings