Learn More
TARGETMOL CHEMICALS INC Rizatriptan benzoate 50MG
SDP
Catalog No. 502878438
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Rizatriptan benzoate (MK-462 Benzoate) selectively binds to and activates serotonin (5-HT) 1B receptors and 5-HT 1D receptors providing relief of migraine headaches. Purity 99%
Catalog No. 50-287-8438 Supplier TARGETMOL CHEMICALS INC Supplier No. T151250MG
Also available in 1 mL, 10 mg, 25 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Rizatriptan benzoate (MK-462 Benzoate) selectively binds to and activates serotonin (5-HT) 1B receptors and 5-HT 1D receptors providing relief of migraine headaches. Purity 99%