Learn More
Cayman Chemical Rizatriptan (benzoate salt), Purity: ≥98%, CAS: 145202-66-0, 50 mg
SDP
Catalog No. 502970962
Encompass_Preferred
A 5-HT1B and 5-HT1D receptor agonist (Kis = 4.3 and 10.1 nM, respectively); selective for 5-HT1B and 5-HT1D receptors over 5-HT1A receptors (Ki = 140 nM); induces vasoconstriction in isolated human middle meningeal arteries (EC50 = 90 nM); reduces pain behaviors and photophobia in a Cacna1a mutant transgenic mouse model of migraine at 50 and 75 mg/kg
Catalog No. 50-297-0962 Supplier Cayman Chemical Supplier No. 1647550 MG
A 5-HT1B and 5-HT1D receptor agonist (Kis = 4.3 and 10.1 nM, respectively); selective for 5-HT1B and 5-HT1D receptors over 5-HT1A receptors (Ki = 140 nM); induces vasoconstriction in isolated human middle meningeal arteries (EC50 = 90 nM); reduces pain behaviors and photophobia in a Cacna1a mutant transgenic mouse model of migraine at 50 and 75 mg/kg