Learn More
Pyramex™ RJ31 Series Winter Wear Bomber Jacket
Catalog No. 19555268 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Hi-vis Lime
Hi-vis Orange
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-555-268 Hi-vis Orange X-Large
19-555-256 Hi-vis Lime Medium
19-555-255 Hi-vis Lime Large
19-555-261 Hi-vis Lime X-Large
19-555-257 Hi-vis Lime 2X-Large
19-555-258 Hi-vis Lime 3X-Large
19-555-259 Hi-vis Lime 4X-Large
19-555-260 Hi-vis Lime 5X-Large
19-555-263 Hi-vis Orange Medium
19-555-262 Hi-vis Orange Large
19-555-264 Hi-vis Orange 2X-Large
19-555-265 Hi-vis Orange 3X-Large
19-555-266 Hi-vis Orange 4X-Large
19-555-267 Hi-vis Orange 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Catalog No. 19-555-268 Supplier Pyramex™ Supplier No. RJ3120XL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Pyramex™ Safety Hi -Vis Winter Wear
  • Weatherproof polyester shell with zip out polar fleece liner.
  • Pockets on outer shell and removable fleece liner. 8 pockets-5 outside, 3 inside.
  • Cell phone/radio chest pocket with clear panel on outside shell.
  • Detachable drawstring hood has collar pouch to hide away hood when not in use.
  • 2 inch silver reflective tape.
  • Two double tacked microphone tabs on left and right shoulders on front of jacket.
Specifications
Size X-Large
Material Weatherproof Polyester
Color Hi-vis Orange
Closure Type Zipper
Certifications/Compliance Meets ANSI/ISEA 107-2015; Type R - Class 3
Coating Polyurethane
Cuff Style Elastic
Hood Type Concealed, Detachable
Liner Material Polar Fleece
No. of Pockets 8
Tape Type China Brand Flat Silver Reflective Tape