Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RJPXD33 | 1613160-77-2 | 98.1% | 1489.78 | 50 MG
SDP

Catalog No. 5000423098
Encompass_Preferred

RJPXD33 is an inhibitor for LpxD and LpxA with IC50 values of 3.5 μM and 19 μM, respectively. It inhibits LpxD and LpxA in *E. coli* with Kd values of 6 μM and 20 μM.

  • Inhibits LpxD and LpxA
  • IC50 values of 3.5 μM (LpxD) and 19 μM (LpxA)
  • Inhibits LpxD and LpxA in *E. coli* with Kd values of 6 μM and 20 μM
  • Appearance: solid
  • Color: white to off-white
  • Sequence: Thr-Asn-Leu-Tyr-Met-Leu-Pro-Lys-Trp-Asp-Ile-Pro-NH2
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month (sealed storage, away from moisture)

Catalog No. 50-004-23098 Supplier Medchemexpress LLC Supplier No. HYP1084850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RJPXD33 is an inhibitor for LpxD and LpxA with IC50 values of 3.5 μM and 19 μM, respectively. It inhibits LpxD and LpxA in *E. coli* with Kd values of 6 μM and 20 μM.

  • Inhibits LpxD and LpxA
  • IC50 values of 3.5 μM (LpxD) and 19 μM (LpxA)
  • Inhibits LpxD and LpxA in *E. coli* with Kd values of 6 μM and 20 μM
  • Appearance: solid
  • Color: white to off-white
  • Sequence: Thr-Asn-Leu-Tyr-Met-Leu-Pro-Lys-Trp-Asp-Ile-Pro-NH2
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month (sealed storage, away from moisture)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.