Learn More
Cayman Chemical RJR 2429 (hydrochloride), Purity: ≥98%, CAS: 1021418-53-0, 10 mg
SDP
Catalog No. 502989228
Encompass_Preferred
An agonist of nicotinic AChRs; selective for α4β2 or α7 subunit-containing nicotinic AChRs (Kis = 0.7 and 10.9 nM in isolated rat cortex and isolated rat hippocampus, respectively) over α1βɣδ subunit-containing nicotinic AChRs (EC50 = 1,000 nM in PC12 cells); induces dopamine release from isolated rat striatal synaptosomes (EC50 = 2.4 nM) and increases nicotine-induced elevations in epinephrine and norepinephrine levels in isolated rat adrenal gland at 100 µM; inhibits nicotine-induced ion flux in isolated rat thalamic synaptosomes (IC50 = 154 nM)
Catalog No. 50-298-9228 Supplier Cayman Chemical Supplier No. 2974510 MG
An agonist of nicotinic AChRs; selective for α4β2 or α7 subunit-containing nicotinic AChRs (Kis = 0.7 and 10.9 nM in isolated rat cortex and isolated rat hippocampus, respectively) over α1βɣδ subunit-containing nicotinic AChRs (EC50 = 1,000 nM in PC12 cells); induces dopamine release from isolated rat striatal synaptosomes (EC50 = 2.4 nM) and increases nicotine-induced elevations in epinephrine and norepinephrine levels in isolated rat adrenal gland at 100 µM; inhibits nicotine-induced ion flux in isolated rat thalamic synaptosomes (IC50 = 154 nM)