Learn More
Pyramex™ RJR34 Series Winter Wear Windbreaker
Catalog No. 19555285 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-555-285 Medium
19-555-284 Large
19-555-290 X-Large
19-555-286 2X-Large
19-555-287 3X-Large
19-555-288 4X-Large
19-555-289 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-555-285 Supplier Pyramex™ Supplier No. RJR3410M
Only null left
Add to Cart
Add to cart
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Pyramex™ Safety Hi -Vis Winter Wear
  • Water resistant 4-in-1 reversible windbreaker polyester jacket
  • Weatherproof elastic band at wrists
  • Removable zip-out sleeves — converts Class 3 jacket to Class 2 vest
  • Reverses from hi-vis lime to gray/black
  • Silver reflective tape on hi-vis side
  • Hi-vis reflective piping on gray/black side
  • Bottom hem adjustment — bungee cord/toggle pull
  • Mic tab on hi-vis side
Specifications
Size Medium
Material Polyester
Color Hi-vis Lime
Closure Type Zipper
Certifications/Compliance Meets ANSI/ISEA 107-2015; Type R - Class 3 with sleeves hi-vis side only; Type R - Class 2 without sleeves hi-vis side only
Coating TPU Coating and Water Resistant Treatment
No. of Pockets 3 per side
Sleeve Style Removable, Zip-out
Tape Type China Brand Silver Reflective Stripes