Learn More
Medchemexpress LLC HY-123892 5mg Medchemexpress, RK-287107 CAS:2171386-10-8 Purity:>98%
SDP
Catalog No. 501932584
Encompass_Preferred
Medchemexpress, HY-123892 5mg RK-287107 CAS:2171386-10-8 RK-287107 is a potent and specific tankyrase inhibitor with IC 50 s of 14.3 and 10.6 nM for tankyrase-1 and tankyrase-2 , respectively. RK-287107 blocks colorectal cancer cell growth [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2584 Supplier Medchemexpress LLC Supplier No. HY1238925MG
Medchemexpress, HY-123892 5mg RK-287107 CAS:2171386-10-8 RK-287107 is a potent and specific tankyrase inhibitor with IC 50 s of 14.3 and 10.6 nM for tankyrase-1 and tankyrase-2 , respectively. RK-287107 blocks colorectal cancer cell growth [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.