Learn More
Selleck Chemical LLC RK-33
SDP
Supplier:  Selleck Chemical LLC S824625MG
Encompass_Preferred
RK-33 is a first-in-class small molecule inhibitor of DDX3 (a RNA helicase) and causes G1 cell cycle arrest induces apoptosis and promotes radiation sensitization in DDX3-overexpressing cells
Catalog No. 50-000-01916