Learn More
Selleck Chemical LLC RK-33 2mg 1070773-09-9
SDP
Catalog No. NC1261730
Encompass_Preferred
RK-33 is a first-in-class small molecule inhibitor of DDX3 (a RNA helicase) and causes G1 cell cycle arrest, induces apoptosis, and promotes radiation sensitization in DDX3-overexpressing cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1261730 Supplier Selleck Chemical LLC Supplier No. S82462MG
RK-33 is a first-in-class small molecule inhibitor of DDX3 (a RNA helicase) and causes G1 cell cycle arrest, induces apoptosis, and promotes radiation sensitization in DDX3-overexpressing cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.