Learn More
Medchemexpress LLC Rk-582 | 2171388-28-4 | 98.05% | 510.60 | 25 MG
SDP
Catalog No. 5000347792
Encompass_Preferred
RK-582 is a tankyrase inhibitor and an orally bioavailable growth inhibitor, acting as an antitumor agent. It is suitable for research on colorectal cancer. This compound potently inhibits human tankyrase-1 and tankyrase-2 catalytic activity with high selectivity over PARP1, PARP2, and PARP10. It also inhibits Wnt/β-catenin signaling and the growth of COLO-320DM human colorectal cancer cells.
  • Inhibits tankyrase-1 and tankyrase-2 catalytic activity
  • Demonstrates high selectivity over PARP1, PARP2, and PARP10
  • Inhibits Wnt/β-catenin signaling
  • Potently inhibits the growth of COLO-320DM human colorectal cancer cells
  • Modulates Wnt/β-catenin signaling by stabilizing AXIN2 and reducing active β-catenin
Catalog No. 50-003-47792 Supplier Medchemexpress LLC Supplier No. 25MGHY137849
RK-582 is a tankyrase inhibitor and an orally bioavailable growth inhibitor, acting as an antitumor agent. It is suitable for research on colorectal cancer. This compound potently inhibits human tankyrase-1 and tankyrase-2 catalytic activity with high selectivity over PARP1, PARP2, and PARP10. It also inhibits Wnt/β-catenin signaling and the growth of COLO-320DM human colorectal cancer cells.
  • Inhibits tankyrase-1 and tankyrase-2 catalytic activity
  • Demonstrates high selectivity over PARP1, PARP2, and PARP10
  • Inhibits Wnt/β-catenin signaling
  • Potently inhibits the growth of COLO-320DM human colorectal cancer cells
  • Modulates Wnt/β-catenin signaling by stabilizing AXIN2 and reducing active β-catenin