Learn More
Medchemexpress LLC RK-582 | 2171388-28-4 | 98.05% | 510.60 | 50 MG
SDP
Catalog No. 5000352221
Encompass_Preferred
RK-582 is a tankyrase inhibitor, antitumor agent, and orally bioavailable growth inhibitor. It has an IC50 of 36.1 nM against human tankyrase-1 and 39.2 nM against human tankyrase-2. In APC-mutated colorectal cancer cells, sensitivity to RK-582 correlates with the level of active β-catenin, while drug resistance is associated with PIK3CA mutation. RK-582 can be used for the research of colorectal cancer.
  • Tankyrase inhibitor
  • Antitumor agent
  • Orally bioavailable growth inhibitor
  • IC50 of 36.1 nM against human tankyrase-1
  • IC50 of 39.2 nM against human tankyrase-2
  • Correlates with active β-catenin levels in APC-mutated colorectal cancer cells
  • Useful for colorectal cancer research
Catalog No. 50-003-52221 Supplier Medchemexpress LLC Supplier No. HY13784950MG
RK-582 is a tankyrase inhibitor, antitumor agent, and orally bioavailable growth inhibitor. It has an IC50 of 36.1 nM against human tankyrase-1 and 39.2 nM against human tankyrase-2. In APC-mutated colorectal cancer cells, sensitivity to RK-582 correlates with the level of active β-catenin, while drug resistance is associated with PIK3CA mutation. RK-582 can be used for the research of colorectal cancer.
  • Tankyrase inhibitor
  • Antitumor agent
  • Orally bioavailable growth inhibitor
  • IC50 of 36.1 nM against human tankyrase-1
  • IC50 of 39.2 nM against human tankyrase-2
  • Correlates with active β-catenin levels in APC-mutated colorectal cancer cells
  • Useful for colorectal cancer research