Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rk-582 | 2171388-28-4 | 98.1% | 510.6 g·mol⁻1 | C27H35FN6O3 | 5 MG
SDP

Catalog No. 5000222875
Encompass_Preferred

RK-582 is an orally active, spiroindolinone-based selective tankyrase inhibitor used as a research reagent to probe Wnt signaling and tumor growth pathways. It exhibits nanomolar potency against tankyrase enzymes and has shown efficacy in preclinical in vivo models. The compound is supplied as a purified solid with recommended storage and solubility guidance for laboratory use.

  • Orally active spiroindolinone-based tankyrase inhibitor.
  • High potency in the nanomolar range against tankyrase enzymes.
  • Demonstrated efficacy in preclinical tumor xenograft models.
  • High purity suitable for biochemical and cellular studies.
  • Soluble in DMSO with established in vivo formulation protocols.
  • Recommended storage conditions provided for long-term and solvent storage.

Catalog No. 50-002-22875 Supplier Medchemexpress LLC Supplier No. HY1378495MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RK-582 is an orally active, spiroindolinone-based selective tankyrase inhibitor used as a research reagent to probe Wnt signaling and tumor growth pathways. It exhibits nanomolar potency against tankyrase enzymes and has shown efficacy in preclinical in vivo models. The compound is supplied as a purified solid with recommended storage and solubility guidance for laboratory use.

  • Orally active spiroindolinone-based tankyrase inhibitor.
  • High potency in the nanomolar range against tankyrase enzymes.
  • Demonstrated efficacy in preclinical tumor xenograft models.
  • High purity suitable for biochemical and cellular studies.
  • Soluble in DMSO with established in vivo formulation protocols.
  • Recommended storage conditions provided for long-term and solvent storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.