Learn More
Enzo Life Sciences RK-682 (200µg). CAS: 154639-24-4
SDP
Catalog No. 502011507
Encompass_Preferred
RK-682 is a novel tyrosine phosphatase inhibitor isolated from Streptomyces sp. It inhibits the dual specificity VHR phosphatase (IC50=2.0µM) selectively over CD45 (IC50=54µM), Cdc25B and PTP1B (IC50~42µM). RK-682 inhibits cell cycle progression of Ball-1 cells at the G1/S transition and enhances ATP-induced long-term potentiation in guinea pig hippocampal CA1 neurons. Shows potent activity against HIV-1 protease. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25mg/ml), 100% ethanol (10mg/ml), methanol or dimethyl formamide. Long Term Storage: -20°C.
Catalog No. 50-201-1507 Supplier Enzo Life Sciences Supplier No. BMLPR1120200
RK-682 is a novel tyrosine phosphatase inhibitor isolated from Streptomyces sp. It inhibits the dual specificity VHR phosphatase (IC50=2.0µM) selectively over CD45 (IC50=54µM), Cdc25B and PTP1B (IC50~42µM). RK-682 inhibits cell cycle progression of Ball-1 cells at the G1/S transition and enhances ATP-induced long-term potentiation in guinea pig hippocampal CA1 neurons. Shows potent activity against HIV-1 protease. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25mg/ml), 100% ethanol (10mg/ml), methanol or dimethyl formamide. Long Term Storage: -20°C.