Learn More
Medchemexpress LLC Rk-701 | 2648855-18-7 | 98.0% | 446.54 | C26H30N4O3 | 10 MG
SDP
Catalog No. 5000286262
Encompass_Preferred
RK-701 is a selective, non-genotoxic small-molecule inhibitor of the histone methyltransferase G9a (EHMT2) developed for research use. It exhibits potent biochemical and cellular activity and includes manufacturer-provided data for solubility, stability, and purity to support experimental planning.
Catalog No. 50-002-86262 Supplier Medchemexpress LLC Supplier No. HY15277510MG
RK-701 is a selective, non-genotoxic small-molecule inhibitor of the histone methyltransferase G9a (EHMT2) developed for research use. It exhibits potent biochemical and cellular activity and includes manufacturer-provided data for solubility, stability, and purity to support experimental planning.