Learn More
Selleck Chemical LLC RK-701-E1607-100MG
SDP
Catalog No. 503135126
Encompass_Preferred
RK-701 is a highly selective potent and low-toxicity inhibitor of histone methyltransferases G9a and GLP1 with IC50 values of 23-27 nM and 53 nM RK-701 treatment induces fetal globin expression in human erythroid cells and in mice and plays an essential role in -globin induction RK-701 is used in the research of sickle cell disease
Catalog No. 50-313-5126 Supplier Selleck Chemical LLC Supplier No. E1607100MG
RK-701 is a highly selective potent and low-toxicity inhibitor of histone methyltransferases G9a and GLP1 with IC50 values of 23-27 nM and 53 nM RK-701 treatment induces fetal globin expression in human erythroid cells and in mice and plays an essential role in -globin induction RK-701 is used in the research of sickle cell disease