Learn More
ABclonal Technology dNTPs (25 mM each)
SDP
Catalog No. 5000136303
Encompass_Preferred
dNTPs (Deoxynucleotide Solution Mix) is an equimolar solution of ultrapure dATP, dCTP, dGTP and dTTP. The solution contains 25 mM each of dATP, dCTP, dGTP and dTTP. It is supplied in ultrapure water as a sodium salt at pH 7.5. Purity is above 99% detected by HPLC.
Catalog No. 50-001-36303 Supplier ABclonal Technology Supplier No. RK20101100UL
dNTPs (Deoxynucleotide Solution Mix) is an equimolar solution of ultrapure dATP, dCTP, dGTP and dTTP. The solution contains 25 mM each of dATP, dCTP, dGTP and dTTP. It is supplied in ultrapure water as a sodium salt at pH 7.5. Purity is above 99% detected by HPLC.