Learn More
ABclonal Technology dNTPs (25 mM each)
Supplier: ABclonal Technology RK201015ML
dNTPs (Deoxynucleotide Solution Mix) is an equimolar solution of ultrapure dATP, dCTP, dGTP and dTTP. The solution contains 25 mM each of dATP, dCTP, dGTP and dTTP. It is supplied in ultrapure water as a sodium salt at pH 7.5. Purity is above 99% detected by HPLC.