Learn More
ABclonal Technology Nuclease-free H2O
SDP
Catalog No. 5000135507
Encompass_Preferred
Ideal for the preparation of reagents and for use in enzymatic reactions. No toxic agents, such as DEPC, are used in production, so as to avoid inhibition in enzymatic reactions. Available in 25 ml size.
Catalog No. 50-001-35507 Supplier ABclonal Technology Supplier No. RK20102100ML
Ideal for the preparation of reagents and for use in enzymatic reactions. No toxic agents, such as DEPC, are used in production, so as to avoid inhibition in enzymatic reactions. Available in 25 ml size.