Learn More
ABclonal Technology dTTP (100 mM)
SDP
Supplier:  ABclonal Technology RK201111ML
Encompass_Preferred
Deoxythymidine triphosphate (dTTP) is one of the four nucleoside triphosphates that are used in the in vivo synthesis of DNA. Anucleoside triphosphate is a molecule type that contains a nucleoside with three phosphates bound to it. It can be used in PCR, RT, qPCR, RT-qPCR, NGS and other molecular biology experiments. The concentration of this product is 100 mM, diluted withultrapure water as a sodium salt at pH 7.5.
Catalog No. 50-001-35664