Learn More
ABclonal Technology dGTP (100 mM)
SDP
Supplier:  ABclonal Technology RK20112100UL
Encompass_Preferred
Deoxyguanosine triphosphate (dGTP) is a nucleoside triphosphate used in cells for DNA synthesis. A nucleoside triphosphateis a molecule type that contains a nucleoside with three phosphates bound to it. It can be used in PCR, RT, qPCR, RT-qPCR, NGS and other molecular biology experiments. The concentration of this product is 100 mM, diluted with ultrapure water as a sodium salt at pH 7.5.
Catalog No. 50-001-35662