Learn More
ABclonal Technology dCTP (100 mM)
SDP
Supplier:  ABclonal Technology RK20113100UL
Encompass_Preferred
Deoxycytidine triphosphate (dCTP) is a nucleoside phosphate in being comprised of a deoxyribonucleoside and three phosphate units. This means that it has a deoxyribose as its sugar constituent with three phosphate units attached. Its nucleoside contains a pyrimidine base. It can be used in PCR, RT, qPCR, RT-qPCR, NGS and other molecular biology experiments. The concentration of this product is 100 mM, diluted with ultrapure water as a sodium salt at pH 7.5.
Catalog No. 50-001-35505