Learn More
ABclonal Technology Oligo d(T)23VN (50 μM)
SDP
Catalog No. 5000136114
Encompass_Preferred
Oligo d(T)23VN is used for the priming and sequencing of mRNA adjacent to the 3-poly A tail. V equal A or G or C N equal A or G or C or T Reconstitute with water.
Catalog No. 50-001-36114 Supplier ABclonal Technology Supplier No. RM20115100UL
Oligo d(T)23VN is used for the priming and sequencing of mRNA adjacent to the 3-poly A tail. V equal A or G or C N equal A or G or C or T Reconstitute with water.