Learn More
ABclonal Technology dNTPs (10 mM each)
SDP
Supplier:  ABclonal Technology RK201205ML
Encompass_Preferred
dNTPs (Deoxynucleotide Solution Mix) is an equimolar solution of ultrapure dATP, dCTP, dGTP and dTTP. The solution contains 10 mM each of dATP, dCTP, dGTP and dTTP. Each vial contains 10 μmol in a volume of 1 ml. A small pack contains 1 vial of 1 ml and a large pack contains 5 vials of 1 ml. Supplied in: Ultrapure water as a sodium salt at pH 7.5.
Catalog No. 50-001-34607