Learn More
ABclonal Technology Damaged DNA Repair Enzymes II
SDP
Supplier:  ABclonal Technology RK2023424RXN
Encompass_Preferred
FFPE refers to formalin-fixed paraffin-embedded samples, in which DNA is severely damaged. Damaged DNA Repair Enzymes II is an optimized enzyme mixture primarily used to repair FFPE sample DNA, thereby obtaining high-quality sequencing data.
Catalog No. 50-001-36285