Learn More
ABclonal Technology AFTMag NGS DNA Clean Beads
SDP
Catalog No. 5000136239
Encompass_Preferred
AFTMag NGS DNA Clean Beads is a SPRI-based chemistry that speeds and simplifies nucleic acid size selection and PCR purification for DNA and/or RNA library preparation for Next Generation sequencing. It is compatible with mainstream NGS library prepared kits. So,it effectively reduce your library preparation costs with the same effect. The protocol can be adapted to your current liquid handling workstation (e.g. Beckman, Hamilton, Tecan, Caliper, Perkin Elmer, Agilent and Eppendorf) utilizing your current protocol as well as it can be performed manually.
Catalog No. 50-001-36239 Supplier ABclonal Technology Supplier No. RK202575ML
AFTMag NGS DNA Clean Beads is a SPRI-based chemistry that speeds and simplifies nucleic acid size selection and PCR purification for DNA and/or RNA library preparation for Next Generation sequencing. It is compatible with mainstream NGS library prepared kits. So,it effectively reduce your library preparation costs with the same effect. The protocol can be adapted to your current liquid handling workstation (e.g. Beckman, Hamilton, Tecan, Caliper, Perkin Elmer, Agilent and Eppendorf) utilizing your current protocol as well as it can be performed manually.