Learn More
ABclonal Technology FS Pro DNA Lib Prep Kit V2
SDP
Catalog No. 5000136240
Encompass_Preferred
The FS Pro DNA Lib Prep Kit for Illumina streamlines NGS library prep for Illumina platforms by combining DNA fragmentation, end repair, and dA-tailing in one step, enabling direct adapter ligation without purification. The entire process takes under 2 hours, supports diverse DNA sources (including FFPE samples), and reduces false positives for improved sequencing yield. Ideal for PCR-free libraries (e.g., 100 ng high-quality gDNA), it undergoes rigorous QC (contamination, functionality, and stability tests) to ensure reliability, including pathogen detection compatibility.
Catalog No. 50-001-36240 Supplier ABclonal Technology Supplier No. RK2027596RXN
The FS Pro DNA Lib Prep Kit for Illumina streamlines NGS library prep for Illumina platforms by combining DNA fragmentation, end repair, and dA-tailing in one step, enabling direct adapter ligation without purification. The entire process takes under 2 hours, supports diverse DNA sources (including FFPE samples), and reduces false positives for improved sequencing yield. Ideal for PCR-free libraries (e.g., 100 ng high-quality gDNA), it undergoes rigorous QC (contamination, functionality, and stability tests) to ensure reliability, including pathogen detection compatibility.