Learn More
ABclonal Technology Second Strand Synthesis Module
SDP
Catalog No. 5000138433
Encompass_Preferred
The ABclonal Second Strand Synthesis Module (RK20346) uses productfrom the ABclonal First Strand cDNA Synthesis Module (RK20342 orRK20353) as a template for second strand cDNA synthesis to generate adouble-stranded cDNA product. This double-stranded cDNA product canbe directly used for DNA library construction.
Catalog No. 50-001-38433 Supplier ABclonal Technology Supplier No. RK2034624RXN
The ABclonal Second Strand Synthesis Module (RK20346) uses productfrom the ABclonal First Strand cDNA Synthesis Module (RK20342 orRK20353) as a template for second strand cDNA synthesis to generate adouble-stranded cDNA product. This double-stranded cDNA product canbe directly used for DNA library construction.