Learn More
ABclonal Technology Small RNA Adapter Module 96 Index for Illumina MidiSet (24 indices)
SDP
Catalog No. 5000137431
Encompass_Preferred
The Small RNA Adapter Module 96 Index for Illumina should be used with the small RNA-seq library preparation kit for library amplification. It contains the sRNA Universal PCR Primer and index primer and can be used during PCR to obtain a small RNA-seq library with an intact structure. All reagents provided in the kit have undergone strict quality control and function validation to ensure the stability and repeatability of library preparation.
Catalog No. 50-001-37431 Supplier ABclonal Technology Supplier No. RK2035824RXN
The Small RNA Adapter Module 96 Index for Illumina should be used with the small RNA-seq library preparation kit for library amplification. It contains the sRNA Universal PCR Primer and index primer and can be used during PCR to obtain a small RNA-seq library with an intact structure. All reagents provided in the kit have undergone strict quality control and function validation to ensure the stability and repeatability of library preparation.