Learn More
ABclonal Technology ABScript II One Step SYBR Green RT-qPCR Kit
SDP
Catalog No. 5000135176
Encompass_Preferred
The ABScript II One-Step SYBR Green RT-qPCR Kit enables efficient one-step RT-qPCR using SYBR Green I fluorescence. It allows reverse transcription and PCR in a single tube, minimizing contamination risks and increasing throughput. Ideal for RNA virus detection, it offers high sensitivity with real-time monitoring, eliminating post-PCR electrophoresis. The kit combines ABScript II Reverse Transcriptase and Taq DNA Polymerase with an optimized buffer for high efficiency and specificity. Ready-to-use enzyme mixes simplify operation. 
Catalog No. 50-001-35176 Supplier ABclonal Technology Supplier No. RK2040425RXN
The ABScript II One-Step SYBR Green RT-qPCR Kit enables efficient one-step RT-qPCR using SYBR Green I fluorescence. It allows reverse transcription and PCR in a single tube, minimizing contamination risks and increasing throughput. Ideal for RNA virus detection, it offers high sensitivity with real-time monitoring, eliminating post-PCR electrophoresis. The kit combines ABScript II Reverse Transcriptase and Taq DNA Polymerase with an optimized buffer for high efficiency and specificity. Ready-to-use enzyme mixes simplify operation. 