Learn More
ABclonal Technology ABScript III Reverse Transcriptase
SDP
Catalog No. 5000135941
Encompass_Preferred
ABScript III Reverse Transcriptase is a genetically engineered M-MuLV reverse transcriptase with reduced RNase H activity and improved thermal stability. Compared with wild-type M-MuLV, it can synthesize first-strand cDNA at higher temperature. Product still has features such as high activity at 50-55℃, higher specificity, higher yield, and the ability to synthesize cDNA up to 12 kb.
Catalog No. 50-001-35941 Supplier ABclonal Technology Supplier No. RK204084000U
ABScript III Reverse Transcriptase is a genetically engineered M-MuLV reverse transcriptase with reduced RNase H activity and improved thermal stability. Compared with wild-type M-MuLV, it can synthesize first-strand cDNA at higher temperature. Product still has features such as high activity at 50-55℃, higher specificity, higher yield, and the ability to synthesize cDNA up to 12 kb.