Learn More
ABclonal Technology ABScript III One Step RT-qPCR Probe Kit with UDG V5
SDP
Catalog No. 5000136621
Encompass_Preferred
ABScript III One Step RT-qPCR Probe Kit with UDG V5 is a universal kit for one step RT-qPCR reactions using the probe method. This kit uses RNA as the template, reverse transcription and PCR reactions can be performed continuously in the same tube, which improves the detection throughput. This kit introduces the dUTP/UDG anti-contamination system, heat-sensitive UDG can rapidly degrade U-containing contaminants at room temperature; and the heat-sensitive UDG is rapidly inactivated during reverse transcription at 50℃, which will not affect the efficiency and sensitivity of RT-qPCR. The reaction system can perform real-time detection of amplified products and improve the detection sensitivity, which is very suitable for the detection of trace RNA such as RNA viruses.
Catalog No. 50-001-36621 Supplier ABclonal Technology Supplier No. RK20412250RXN
ABScript III One Step RT-qPCR Probe Kit with UDG V5 is a universal kit for one step RT-qPCR reactions using the probe method. This kit uses RNA as the template, reverse transcription and PCR reactions can be performed continuously in the same tube, which improves the detection throughput. This kit introduces the dUTP/UDG anti-contamination system, heat-sensitive UDG can rapidly degrade U-containing contaminants at room temperature; and the heat-sensitive UDG is rapidly inactivated during reverse transcription at 50℃, which will not affect the efficiency and sensitivity of RT-qPCR. The reaction system can perform real-time detection of amplified products and improve the detection sensitivity, which is very suitable for the detection of trace RNA such as RNA viruses.