Learn More
ABclonal Technology E.coli DNA Ligase
SDP
Catalog No. 5000136490
Encompass_Preferred
E. coli DNA Ligase catalyzes the formation of a phosphodiester bond between the 5-phosphate and the 3-hydroxyl of two adjacent DNA strands in duplex DNA with cohesive ends. It is not appreciably active on blunt-ended substrates. E. coli DNA Ligase uses NAD as a cofactor and can be heat-inactivated. E. coli DNA Ligase is active at a range of temperatures (4°C - 37°C).
Catalog No. 50-001-36490 Supplier ABclonal Technology Supplier No. RK20501200U
E. coli DNA Ligase catalyzes the formation of a phosphodiester bond between the 5-phosphate and the 3-hydroxyl of two adjacent DNA strands in duplex DNA with cohesive ends. It is not appreciably active on blunt-ended substrates. E. coli DNA Ligase uses NAD as a cofactor and can be heat-inactivated. E. coli DNA Ligase is active at a range of temperatures (4°C - 37°C).