Learn More
ABclonal Technology T4 RNA Ligase 1
SDP
Catalog No. 5000136623
Encompass_Preferred
T4 RNA ligase 1 catalyzes the formation of a 3 to 5 phosphodiester bond by joining the 5 phosphate end of a nucleotide to the 3 hydroxyl end, accompanied by the hydrolysis of ATP to AMP and PPi. Substrates include single-stranded RNA, DNA, and dinucleoside pyrophosphates.
Catalog No. 50-001-36623 Supplier ABclonal Technology Supplier No. RK205031000U
T4 RNA ligase 1 catalyzes the formation of a 3 to 5 phosphodiester bond by joining the 5 phosphate end of a nucleotide to the 3 hydroxyl end, accompanied by the hydrolysis of ATP to AMP and PPi. Substrates include single-stranded RNA, DNA, and dinucleoside pyrophosphates.