Learn More
ABclonal Technology T4 RNA Ligase 2, truncated KQ
SDP
Catalog No. 5000135895
Encompass_Preferred
T4 RNA Ligase 2, truncated KQ (T4 Rnl2tr R55K, K227Q), specifically catalyzes the ligation of a 5-adenylated DNA or RNA to the 3 OH end of an RNA molecule. This enzyme-mediated ligation does not require ATP but necessitates a pre-adenylated adapter. T4 RNA Ligase 2, truncated KQ, a double-point mutant of T4 RNA Ligase 2 with R55K and K227Q substitutions, significantly reduces the non-specific ligation of RNA (such as concatenation or self-circularization) compared to T4 RNA Ligase 2, truncated, while maintaining enzyme activity similar to that of T4 RNA Ligase 2, truncated.
Catalog No. 50-001-35895 Supplier ABclonal Technology Supplier No. RK2050410000U
T4 RNA Ligase 2, truncated KQ (T4 Rnl2tr R55K, K227Q), specifically catalyzes the ligation of a 5-adenylated DNA or RNA to the 3 OH end of an RNA molecule. This enzyme-mediated ligation does not require ATP but necessitates a pre-adenylated adapter. T4 RNA Ligase 2, truncated KQ, a double-point mutant of T4 RNA Ligase 2 with R55K and K227Q substitutions, significantly reduces the non-specific ligation of RNA (such as concatenation or self-circularization) compared to T4 RNA Ligase 2, truncated, while maintaining enzyme activity similar to that of T4 RNA Ligase 2, truncated.