Learn More
ABclonal Technology Circle Ligase
Supplier: ABclonal Technology RK205085000U
Circle Ligase is a thermostable ATP-dependent ligase that can catalyze the self-circularization of single-stranded DNA (ssDNA)/single-stranded RNA (ssRNA) strands simultaneously possessing a 5-phosphate group and a 3-hydroxyl group in the absence of complementary sequences, with the length of single-stranded DNA (ssDNA)/single-stranded RNA (ssRNA) being greater than 15bp.