Learn More
ABclonal Technology T3 DNA Ligase
SDP
Catalog No. 5000135179
Encompass_Preferred
T3 DNA Ligase is an ATP-dependent ds DNA ligase from bacteriophage T3. It can catalyze the formation of a phosphodiester bond between juxtaposed 5 phosphate and 3 hydroxyl termini in duplex DNA or RNA. T3 DNA Ligase will join blunt end and cohesive end termini. It and can also repair single-stranded nicks in duplex DNA, RNA or DNA/RNA hybrids and close the gaps in these DNA substrates. In addition, T3 DNA Ligase is more tolerant to NaCl than T4 DNA Ligase (2-fold). As with T4 DNA Ligase , addition of PEG 6000 to the T3 DNA Ligase reaction system can improve the ligation efficiency of the blunt end. 1X T4 DNA Ligase Reaction Buffer can be used in some experiments where PEG 6000 is not available, but the activity of T3 DNA Ligase is reduced to 1/10. In applications where high concentrations of NaCl need to be maintained, we recommend the use of reaction buffers without PEG 6000.
Catalog No. 50-001-35179 Supplier ABclonal Technology Supplier No. RK20509750000U
T3 DNA Ligase is an ATP-dependent ds DNA ligase from bacteriophage T3. It can catalyze the formation of a phosphodiester bond between juxtaposed 5 phosphate and 3 hydroxyl termini in duplex DNA or RNA. T3 DNA Ligase will join blunt end and cohesive end termini. It and can also repair single-stranded nicks in duplex DNA, RNA or DNA/RNA hybrids and close the gaps in these DNA substrates. In addition, T3 DNA Ligase is more tolerant to NaCl than T4 DNA Ligase (2-fold). As with T4 DNA Ligase , addition of PEG 6000 to the T3 DNA Ligase reaction system can improve the ligation efficiency of the blunt end. 1X T4 DNA Ligase Reaction Buffer can be used in some experiments where PEG 6000 is not available, but the activity of T3 DNA Ligase is reduced to 1/10. In applications where high concentrations of NaCl need to be maintained, we recommend the use of reaction buffers without PEG 6000.